Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

CRM37883

Supelco

1,4-Dimethylnaphthalene

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H6(CH3)2
CAS Number:
Molecular Weight:
156.22
Beilstein:
2039842
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

refractive index

n20/D 1.613 (lit.)

bp

262-264 °C/751 mmHg (lit.)

mp

−18 °C (lit.)

density

1.016 g/mL at 25 °C (lit.)

format

neat

storage temp.

2-8°C

SMILES string

Cc1ccc(C)c2ccccc12

InChI

1S/C12H12/c1-9-7-8-10(2)12-6-4-3-5-11(9)12/h3-8H,1-2H3

InChI key

APQSQLNWAIULLK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ. Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate. Download your certificate at: http://www.sigma-aldrich.com

Application

1,4-Dimethylnaphtalene is a growth inhibitor used to enhance dormancy and retard sprouting in harvested potatoes. It has a very low aqueous solubility and is highly volatile. As per the regulation (EC) No 1107/2009 it is approved for use as plant protection product in the European Union. Maximum residuel levels (MRLs) have been set for 1,4-dimethylnaphthalene according Reg. (EU) No 2022/1346 for various products of plant and animal origin from 0.05 - 15 mg/kg. The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Environmental

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service