Skip to Content
MilliporeSigma
All Photos(1)

Documents

175439

Sigma-Aldrich

4-Isopropylaniline

99%

Synonym(s):

p-Aminocumene, Cumidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHC6H4NH2
CAS Number:
Molecular Weight:
135.21
Beilstein:
1930544
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.543 (lit.)

bp

226-227 °C/745 mmHg (lit.)

solubility

H2O: insoluble

density

0.989 g/mL at 25 °C (lit.)

SMILES string

CC(C)c1ccc(N)cc1

InChI

1S/C9H13N/c1-7(2)8-3-5-9(10)6-4-8/h3-7H,10H2,1-2H3

InChI key

LRTFPLFDLJYEKT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-isopropylaniline is a metabolite of phenylurea herbicide isoproturon.

Application

4-Isopropylaniline was used in the hydroamination of phenylacetylene.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

197.6 °F - closed cup

Flash Point(C)

92 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annelies Peeters et al.
Chemical communications (Cambridge, England), 47(14), 4114-4116 (2011-03-08)
Double metal cyanides (DMCs) are highly active recyclable heterogeneous catalysts for hydroamination of phenylacetylene with 4-isopropylaniline. The best hydroamination yields are obtained with Zn-Co DMCs, especially if the particle size is decreased by a reverse emulsion synthesis technique.
Sebastian R Sørensen et al.
Pest management science, 59(10), 1118-1124 (2003-10-17)
Mineralisation of the phenylurea herbicide isoproturon (3-(4-isopropylphenyl)-1,1-dimethylurea) and two of its known metabolites, 3-(4-isopropylphenyl)-1-methylurea (monodesmethyl-isoproturon) and 4-isopropylaniline, was studied in Danish agricultural soils with or without previous exposure to isoproturon. A potential for rapid mineralisation of isoproturon and the two
Luka A Wright et al.
Dalton transactions (Cambridge, England : 2003), 44(16), 7230-7241 (2015-03-20)
The 2-(2′-aniline)-6-imine-pyridines, 2-(C6H4-2′-NH2)-6-(CMe=NAr)C5H3N (Ar = 4-i-PrC6H4 (HL1a), 2,6-i-Pr2C6H3 (HL1b)), have been synthesised via sequential Stille cross-coupling, deprotection and condensation steps from 6-tributylstannyl-2-(2-methyl-1,3-dioxolan-2-yl)pyridine and 2-bromonitrobenzene. The palladium(II) acetate N,N,N-pincer complexes, [{2-(C6H4-2′-NH)-6-(CMe=NAr)C5H3N}Pd(OAc)] (Ar = 4-i-PrC6H4 (1a), 2,6-i-Pr2C6H3 (1b)), can be prepared by
Luka A Wright et al.
Dalton transactions (Cambridge, England : 2003), 44(13), 6040-6051 (2015-02-28)
The syntheses of two families of sterically tuneable O,N,N pro-ligands are reported, namely the 2-(phenyl-2'-ol)-6-imine-pyridines, 2-(C6H4-2'-OH),6-(CMe=NAr)C5H3N [Ar = 4-i-PrC6H4 (HL1(a)), 2,6-i-Pr2C6H3 (HL1(b))] and the 2-(phenyl-2'-ol)-6-(amino-prop-2-yl)pyridines, 2-(C6H4-2'-OH),6-(CMe2NHAr)C5H3N [Ar = 4-i-PrC6H4 (HL2(a)), 2,6-i-Pr2C6H3 (HL2(b))], using straightforward synthetic approaches and in reasonable overall

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service