Skip to Content
MilliporeSigma
All Photos(1)

Documents

PHR1127

Supelco

Amoxicillin trihydrate

Pharmaceutical Secondary Standard; Certified Reference Material

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O5S · 3H2O
CAS Number:
Molecular Weight:
419.45
Beilstein:
7507120
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP 19
traceable to Ph. Eur. A0800000
traceable to USP 1031503

API family

amoxicillin

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

-10 to -25°C

SMILES string

O.O.O.CC1(C)S[C@@H]2[C@H](NC(=O)[C@H](N)c3ccc(O)cc3)C(=O)N2[C@H]1C(O)=O

InChI

1S/C16H19N3O5S.3H2O/c1-16(2)11(15(23)24)19-13(22)10(14(19)25-16)18-12(21)9(17)7-3-5-8(20)6-4-7;;;/h3-6,9-11,14,20H,17H2,1-2H3,(H,18,21)(H,23,24);3*1H2/t9-,10-,11+,14-;;;/m1.../s1

InChI key

MQXQVCLAUDMCEF-CWLIKTDRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Amoxicillin trihydrate is a oral semisynthetic penicillin, which shows antibacterial activity against Gram positive and and a limited range of Gram-negative organisms. Its mode of action involves the inhibition of bacterial cell wall synthesis.

Application

Amoxicillin trihydrate may be used pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using liquid chromatography technique and spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAB3723 in the slot below. This is an example certificate only and may not be the lot that you receive.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectrophotometric determination of amoxicillin in pharmaceutical formulations
Unal K, et al.
Turkish Journal of Pharmaceutical Sciences, 5(1), 1-16 (2008)
Validated stability-indicating LC method for estimation of amoxicillin trihydrate in pharmaceutical dosage forms and time-dependent release formulations
Beg S, et al.
International Journal of Pharmaceutical Sciences and Nanotechnology, 4(2) (2011)
Adriaan Blommaert et al.
The Journal of antimicrobial chemotherapy, 68(10), 2383-2392 (2013-05-18)
Bacterial resistance to antibiotics, driven by antibiotic consumption, imposes a major threat to the effective treatment of bacterial infections. In addition to reducing the amount of antibiotics prescribed, avoiding broad-spectrum antibiotics could extend the lifetime of the current arsenal of
Aluma Chovel-Sella et al.
Pediatrics, 131(5), e1424-e1427 (2013-04-17)
"Ampicillin rash," a phenomenon unique to patients with Epstein-Barr virus acute infectious mononucleosis (AIM) treated with ampicillin, was first reported in the 1960s. The incidence was estimated as being between 80% and 100%, and the figures have not been reviewed
Allan S Lieberthal et al.
Pediatrics, 131(3), e964-e999 (2013-02-27)
This evidence-based clinical practice guideline is a revision of the 2004 acute otitis media (AOM) guideline from the American Academy of Pediatrics (AAP) and American Academy of Family Physicians. It provides recommendations to primary care clinicians for the management of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service