Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

818674

TSKgel® SuperSW2000 (4 µm) HPLC Columns

L x I.D. 30 cm × 4.6 mm, diol phase

Synonym(s):

TSKgel® Size Exclusion (SW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

TSKgel® SuperSW2000 HPLC Column, phase diol, L × I.D. 30 cm × 4.6 mm, 4 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 18674

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
0.4 mL/min flow rate
10-30 °C temperature
120 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 4.6 mm

matrix

spherical silica

matrix active group

diol phase

particle size

4 μm

pore size

125 Å pore diameter

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Featuring ultra-efficient 4μm particles, TSKgel SuperSW2000 provides higher efficiency than 5μm TSKgel G2000SWxl columns. The 125Å pore size results in a fractionation range up to 150,000 Da for globular proteins. The narrow 4.6mm column inner diameter reduces buffer consumption and is ideal for sample-limited applications. To fully benefit from the high efficiency of 4μm columns, and because the column diameter of TSKgel SuperSW columns is 4.6mm, it is important to minimize the amount of dead volume in the HPLC system. Please call tech service if you need assistance.

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service