Skip to Content
MilliporeSigma
All Photos(1)

Documents

808021

TSKgel® G3000PWXL (7 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Size Exclusion (PW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

product name

TSKgel® G3000PWXL HPLC Column, phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle size

material

stainless steel

Agency

suitable for USP L37
suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 08021

parameter

≤0.5 M salt concentration
≤20% organic solvent
1 mL/min flow rate
40 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

7 μm

pore size

200 Å pore diameter

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Use TSKgel® G3000PWXL columns for the analysis of water-soluble linear polymers with molecular weights up to 50,000 Da. The smaller particle size of PWXL-type columns provide 1.7 times higher resolution than their PW counterpart, making PWXL-type columns more suitable for analytical purposes.

Application

TSKgel® G3000PWXL HPLC Column has been used for the separation of:
  • Sea cucumber Apostichopus japonicus (AJP) from marine aquaculture farms during its analysis using high performance gel permeation chromatography (HPGPC).
  • Natural organic matter (NOM) in water during its separation using ultra-performance liquid chromatography (UPLC) combined with electrospray ionization-triple quadruple mass spectrometry (ESI-tQMS).
  • Galactans derived from potato pectin during their analysis using GPC.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiple approaches to assess pectin binding to galectin-3.
Zhang T, et al.
International Journal of Biological Macromolecules, 91(19), 994-1001 (2016)
Antioxidant and antihyperlipidemic activities of polysaccharides from sea cucumber Apostichopus japonicus.
Liu X, et al.
Carbohydrate Polymers, 90(4), 1664-1670 (2012)
Formation of halogenated organic byproducts during medium-pressure UV and chlorine coexposure of model compounds, NOM and bromide.
Zhao Q, et al.
Water Research, 45(19), 6545-6554 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service