Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

808543

TSKgel® SWXL (7 µm) HPLC Columns

L x I.D. 4 cm × 6 mm, diol phase

Synonym(s):

TSKgel® SWXL Columns

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501

Product Name

TSKgel® SWXL Type Guard Column, phase diol, L × I.D. 4 cm × 6 mm, 7 μm particle size

material

stainless steel

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 08543

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1.2 mL/min flow rate
10-30 °C temperature

technique(s)

HPLC: suitable

L × I.D.

4 cm × 6 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

7 μm

pore size

250 Å

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Géraldine A Kok-Jacon et al.
Transgenic research, 14(4), 385-395 (2005-10-06)
The production of dextran in potato tubers and its effect on starch biosynthesis were investigated. The mature dextransucrase (DsrS) gene from Leuconostoc mesenteroides was fused to the chloroplastic ferredoxin signal peptide (FD) enabling amyloplast entry, which was driven by the
Study of the Heat-Treated Human Albumin Stabilization by Caprylate and Acetyltryptophanate.
Kamran Mousavi Hosseini, et al.
Iranian Biomedical Journal, 6, 135-140 (2002)
Ike Jeon
Analyzing Food for Nutrition Labeling and Hazardous Contaminants , 95-95 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service