Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

820026

TSKgel® SWXL (5 µm) HPLC Columns

L x I.D. 30 cm × 7.8 mm, diol phase

Synonym(s):

TSKgel® BioAssist® G3SWXL HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

Product Name

TSKgel® SWXL Columns, phase diol, L × I.D. 30 cm × 7.8 mm, 5 μm particle size, PEEK hardware

material

PEEK hardware

Agency

suitable for USP L20
suitable for USP L59

product line

TSKgel®

manufacturer/tradename

Tosoh 20026

parameter

≤0.5 M salt concentration
0-100% aqueous soluble organic solvents
1.2 mL/min flow rate
10-30 °C temperature
72 bar max. pressure

technique(s)

HPLC: suitable

L × I.D.

30 cm × 7.8 mm

matrix

silica particle platform

matrix active group

diol phase

particle size

5 μm

pore size

250 Å pore diameter

operating pH

2.5-7.5

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

BioAssist G2SWxl, G3SWxl and G4Swxl are made of PEEK housing material to reduce sample absorption to stainless steel or glass. These columns have the same attributes as the corresponding stainless steel columns (TSKgel G2000SWxl, G3000Swxl and G4000SWxl). Widely considered the premium columns for analyzing biopolymers by GFC, TSKgel G3000SWxl columns feature high pore volume per unit column volume, low sample adsorption and excellent column efficiency, all contributing to unsurpassed sample resolution. With 5μm particles and 250Å pores, TSKgel G3000SWxl is an excellent choice for protein and large peptide separations. The fractionation range is applicable for globular protein samples up to 0.5 million Da.

Application

Calibration range: 10,000-500,000 Da (globular proteins)

Physical form

Shipped in 0.05% NaN3 and 0.1 M Na2SO4 in 0.1 M phosphate buffer, pH 6.7.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

BioAssist is a registered trademark of Tosoh Corporation
TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service