Skip to Content
MilliporeSigma
All Photos(1)

Documents

713-0940

HPLC Analysis of Peroxidase on TSKgel® G3000SWXL

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: TSKgel G3000SWXL, 30 cm × 7.8 mm I.D., 5 μm particles (808541)
mobile phase: 0.3M sodium chloride/0.05M phosphate buffer, pH 7.0
flow rate: 1 mL/min
column temp.: 25 °C
detector: UV, 220 nm
injection: 100 μL crude peroxidase preparation from Japanese horseradish (0.15mg)

suitability

application for HPLC

application(s)

life science and biopharma

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Analyte

Description

    Peroxidase

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service