Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

1.06346

Supelco

Sodium dihydrogen phosphate monohydrate

for analysis EMSURE® ACS,Reag. Ph Eur

Synonym(s):

Sodium phosphate monobasic monohydrate, Monosodium phosphate, Sodium dihydrogen phosphate monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4 · H2O
CAS Number:
Molecular Weight:
137.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-449-2
NACRES:
NA.21
grade:
ACS reagent
form:
crystals
solubility:
850 g/L

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

product line

EMSURE®

Assay

99.0-102.0% (alkalimetric)

form

crystals

potency

>2000 mg/kg LD50, oral (Rat)
>7940 mg/kg LD50, skin (Rabbit)

impurities

≤0.01% Insoluble matter

pH

4.1-4.5 (25 °C, 50 g/L in H2O)

solubility

850 g/L

density

2.04 g/cm3 at 20 °C

bulk density

880 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.005%
Fe: ≤0.0005%
K: ≤0.005%
N: ≤0.001%
heavy metals: ≤0.0005%

UV absorption

λ: 280 nm Amax: ≤0.05

storage temp.

2-30°C

SMILES string

[Na+].[H]O[H].OP(O)([O-])=O

InChI

1S/Na.H3O4P.H2O/c;1-5(2,3)4;/h;(H3,1,2,3,4);1H2/q+1;;/p-1

InChI key

BBMHARZCALWXSL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application


  • Development of a multiparticulate drug delivery system for in situ amorphisation.: This study explores the use of sodium dihydrogen phosphate monohydrate in developing a multiparticulate drug delivery system. The research highlights the compound′s role in achieving in situ amorphisation, enhancing drug solubility and bioavailability (Holm TP et al., 2022).

  • Metabolomic characterization of rhubarb species by capillary electrophoresis and ultra-high-pressure liquid chromatography.: Sodium dihydrogen phosphate monohydrate is utilized in the buffer system for capillary electrophoresis and UHPLC, aiding in the metabolomic analysis of rhubarb species. This research underscores its importance in improving separation efficiency and resolution in analytical methods (Tseng YJ et al., 2013).

Linkage

Replaces: SX0709; SX0709-5; SX0710; SX0710-1; SX0710-3; SX0710-5; SX0710-20

Analysis Note

Assay (alkalimetric): 99.0 - 102.0 %
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water, 25 °C): 4.1 - 4.5
UV absorption (280 nm; 1 mol/l; 1 cm; water): ≤ 0.05
Chloride (Cl): ≤ 0.0005 %
Sulfate (SO₄): ≤ 0.003 %
Total nitrogen (N): ≤ 0.001 %
Heavy metals (ACS): ≤ 0.0005 %
Ca (Calcium): ≤ 0.005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.005 %
Corresponds to ACS,Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia E Schantl et al.
Nature communications, 11(1), 721-721 (2020-02-07)
Myo-inositol hexakisphosphate (IP6) is a natural product known to inhibit vascular calcification (VC), but with limited potency and low plasma exposure following bolus administration. Here we report the design of a series of inositol phosphate analogs as crystallization inhibitors, among

Articles

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

Urea in water and wastewater, enzymatic determination - Photometric determination using the indophenol blue method after cleavage of the urea

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service