Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

505361-U

Supelco

LpDNPH S10L solid phase extraction (SPE) Cartridge

bed wt. 350 mg, volume 1 mL, pkg of 50 ea

Synonym(s):

LpDNPH S10L Cartridge

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

Product Name

LpDNPH S10L Cartridge, bed wt. 350 mg, volume 1 mL, pkg of 50 ea

Agency

ASTM® D5197
EPA 100,IP-6A,TO-11A,TO-11A
NIOSH 2016

composition

bed wt., 350 mg
2 4-DNPH (High-purity silica gel)

packaging

pkg of 50 ea

technique(s)

active air sampling: suitable

volume

1 mL

application(s)

air monitoring
environmental
industrial hygiene

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

LpDNPH S10L Cartridge is employed for trapping carbonyls on a high purity silica adsorbent coated with 2,4-dinitrophenylhydrazine, then converted to hydrazone derivatives. The derivatives are eluted from the cartridge in acetonitrile and are analyzed by HPLC.
The S10L cartridges features a reversible design and is for analysts who prefer shorter dimensions and does not require an adaptor for sampling. The cartridge is eluted by connecting to a syringe barrel (empty SPE cartridge) that acts as a reservoir for gravity-fed elution solvent. Meets EPA TO-11A requirements.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service