Skip to Content
MilliporeSigma
All Photos(1)

Documents

54078-U

Supelco

LpDNPH Ozone Scrubber

Rezorian Cartridge, Potassium Iodide (KI) 1.5 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NB.24

product name

LpDNPH Ozone Scrubber, Rezorian Cartridge, Potassium Iodide (KI) 1.5 g

Agency

ASTM® D5197
EPA 100,IP-6A,TO-11A,TO-11A
NIOSH 2016

composition

Potassium Iodide (KI), 1.5 g

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Ozone scrubbers are recommended for sampling carbonyls in high humidity environments. These cartridges contain 1.5 g of high purity potassium iodide (KI) and are available in two cartridge styles. KI traps the ozone which causes a negative formaldehyde interference in DNPH coated devices. The reversible cartridge offers a slip luer end-fitting, and the rezorian cartridge offers a luer lock end-fitting, to enable you to connect your cartridge of choice directly to the inlet of any DNPH cartridge with a like end-fitting. Testing (200 ppb ozone, 50 % RH, 25°C) indicates the ozone scrubber to have an ozone capacity of 100,000 ppb/hr.

Application

LpDNPH Ozone Scrubber may be used as a sampling cartridge for the determination of volatile organic compounds in air samples using gas chromatography technique.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of volatile organic compounds in the atmosphere and their influence on ozone formation
Skybova M, et al.
Fresenius Environmental Bulletin, 15(12B), 1616-1616 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service