Skip to Content
MilliporeSigma
All Photos(1)

Documents

CRM4M7285

Supelco

TO11/IP-6A Aldehyde/Ketone-DNPH Mix

certified reference material, 15 μg/mL each component in acetonitrile (as the aldehyde or ketone), ampule of 3 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 3 × 1 mL

concentration

15 μg/mL each component in acetonitrile (as the aldehyde or ketone)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
TO11/IP-6A Aldehyde/Ketone-DNPH Mix has been used as certified reference material:
  • For the determination of air-dispersed formaldehyde and carbonyl compounds (CCs) as vapors by denuder-filter sampling and gas chromatography method of analysis.
  • To investigate the impact of NO, NO2 and CO in air samples on the capture and retention of formaldehyde and acetaldehyde with the DNPH-cartridge. The reaction products and chromatographic interferences are identified using chromatography techniques.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acetaldehyde-2,4-dinitrophenylhydrazone
    Acetone-2,4-dinitrophenylhydrazone
    Acrolein-2,4-dinitrophenylhydrazone
    Benzaldehyde-2,4-dinitrophenylhydrazone
    Butyraldehyde-2,4-dinitrophenylhydrazone
    Crotonaldehyde-2,4-dinitrophenylhydrazone
    2,5-Dimethylbenzaldehyde 2,4-dinitrophenylhydrazone
    Formaldehyde-2,4-dinitrophenylhydrazone
    Hexaldehyde-2,4-dinitrophenylhydrazone
    Isovaleraldehyde 2,4-dinitrophenylhydrazone
    Propionaldehyde-2,4-dinitrophenylhydrazone
    o-Tolualdehyde 2,4-dinitrophenylhydrazone
    m-Tolualdehyde 2,4-dinitrophenylhydrazone
    p-Tolualdehyde 2,4-dinitrophenylhydrazone
    Valeraldehyde-2,4-dinitrophenylhydrazone
    See All (15)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of the influence of NO2, NO and CO gases on the determination of formaldehyde and acetaldehyde using the DNPH method as applied to polluted environments
Williams J, et al.
Atmospheric Environment, 218(11), 117019-117019 (2019)
Monitoring of Air-Dispersed Formaldehyde and Carbonyl Compounds as Vapors and Adsorbed on Particulate Matter by Denuder-Filter Sampling and Gas Chromatographic Analysis
Dugheri S, et al.
International Journal of Environmental Research and Public Health, 16(11), 1969-1969 (2019)
Stefano Dugheri et al.
International journal of environmental research and public health, 16(11) (2019-06-06)
Carbonyl compounds (CCs) are products present both as vapors and as condensed species adsorbed on the carbonaceous particle matter dispersed in the air of urban areas, due to vehicular traffic and human activities. Chronic exposure to CCs is a potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service