Skip to Content
MilliporeSigma
All Photos(1)

Documents

28221-U

Supelco

DSD-DNPH Diffusive Sampling Device

pkg of 10 ea

Synonym(s):

Passive DNPH sampler

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

Agency

OSHA 1007

packaging

pkg of 10 ea

technique(s)

passive air sampling: suitable

matrix

(diffusive membrane w/ 2,4-DNPH coated silica)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for sampling aldehydes

Application

DSD-DNPH Diffusive sampling device may be used as a passive air sampler for the analysis of volatile organic compounds in air samples for investigating the factors affecting sick building syndrome (SBS) in residential dwellings.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Symptom definitions for SBS (sick building syndrome) in residential dwellings.
Wang LB, et al.
International Journal of Hygiene and Environmental Health, 211(1-2), 114-120 (2008)

Related Content

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service