Key Documents
33304
Mininert® Valve, screw thread
for use with 24/400 mm thread, pkg of 12 ea
About This Item
UNSPSC Code:
41122001
eCl@ss:
32060101
Pricing and availability is not currently available.
material
white PTFE valve
packaging
pkg of 12 ea
application(s)
environmental
compatibility
for use with 24/400 mm thread
General description
These valves are very easy to use.
1. Push the green button to open the valve
2. Insert the needle through the septum and take sample
3. Remove the needle and push to close
Benefits include:
  • PTFE construction for inertness. PTFE is useful when working when acids and organic solvents.
  • Good sealing ability for containing most volalite liquids and gases at low pressures
  • Mininert valves can be used at temperatures up to 105 °F.
  • 0.036 in. (0.9 mm) opening accepts up to 20-gauge needle
Silicone septa, red
Application
Mininert valves are leak-tight closures for vials and other containers. They are available in both crimp and screw thread versions. Only PTFE and glass are in contact with the contents of the container. They are the ideal closure for the following applications:
  • calibration standards
  • air- or moisture-sensitive fluids
  • derivatizating reagents
  • volatile chemicals
Mininert® valve may be used to seal screw-thread side-arm holding Erlenmeyer flasks containing enrichment cultures; during enrichment culture technique which was used to isolate bacteria from activated sludge, capable of degrading a wide range of aromatic hydrocarbons in mixtures, like ethylbenzene (EBZ), o-, m-, and p-xylene, naphthalene, isopropylbenzene (IPB), styrene (STYR), 1,2,4- and 1,3,5-trimethylbenzene (TMB), 2-chlorotoluene, and 1,2-, 1,3-, and 1,4-dichlorobenzene (DCB), as sources of carbon and energy.[1]
Other Notes
The maximum temperature that Mininert Valves can be used at is 105° F. When Mininert Valves are used at high temperatures, returning them to room temperature may cause the valve to leak slightly.
Legal Information
Mininert is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Joseph G Leahy et al.
FEMS microbiology ecology, 43(2), 271-276 (2003-03-01)
The well-characterized toluene-oxidizing bacteria Pseudomonas putida PaW1, P. putida F1, P. mendocina KR1, Burkholderia cepacia G4, B. cepacia JS150, and Ralstonia pickettii PKO1, as well as several strains of bacteria isolated from activated sludge, were examined for their ability to
Tao Liu et al.
Scientific reports, 7(1), 12913-12913 (2017-10-12)
Phosphine (PH