Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

22145-U

Supelco

Gas Sampling Bulb

size 1000 mL, PTFE stopcock

Synonym(s):

Gas Sampling Bulb, 1000 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

PTFE stopcock

Quality Level

technique(s)

whole air sampling: suitable

size

1000 mL

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Use these glass bulbs to trap and transport a gas sample, then remove an aliquot (through the plug-type septum) for analysis. Oven annealed to resist damage during transportation and on-site use. Available with PTFE or high vacuum glass stopcocks. One half-hole cylindrical Thermogreen septum is included with each bulb.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service