Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27003

Supelco

Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100

volume 15 mL, amber glass vial, O.D. × H 21 mm × 70 mm, thread for 18-400

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

F217/PTFE cap liner
PTFE liner
amber glass vial
green melamine resin cap (solid-top)

feature

closure type green melamine solid-top cap
closure type screw top vial

packaging

pkg of 100 ea

O.D. × H

21 mm × 70 mm

volume

15 mL

fitting

thread for 18-400

Looking for similar products? Visit Product Comparison Guide

Application

It was used in decanting serum for residue analysis using GC/MS/MS. It was also used as reaction and development chamber in examining reaction of dioxane solution of oxalyl chloride with an electropositive metal such as Zn and Mg.

Kit Components Also Available Separately

Product No.
Description
SDS

  • 27088-UVials, screw top, amber glass (vial only), volume 15 mL, amber glass vial, thread for 18-400, pkg of 100 eaSDS

  • 27163Screw cap, solid top with PTFE liner, pkg 100, green melamine resin solid cap, F217/PTFE liner, for use with 15 mL vial with 18 mm threadSDS


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human maternal and umbilical cord blood concentrations of polybrominated diphenyl ethers.
Foster, Warren G., et al.
Chemosphere, 84.10, 1301-1309 (2011)
Zn-Mediated Reduction of Oxalyl Chloride Forming CO and Its Application in Carbonylation Reactions.
Markovic?, Martin, et al.
Organic Letters, 17.22, 5618-5621 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service