Skip to Content
MilliporeSigma
All Photos(1)

Documents

27004

Supelco

Vials, screw top with solid green Thermoset cap with PTFE liner, preassembled, pkg of 100

volume 22 mL, amber glass vial, O.D. × H 23 mm × 85 mm, thread for 20-400

Synonym(s):

PK100 22ML AMBER VIAL,SCREW TOP, SOLID CAP W/PTFE LINER

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

F217/PTFE cap liner
PTFE liner
amber glass vial
green melamine resin cap (solid-top)

feature

closure type green melamine solid-top cap
closure type screw top vial

packaging

pkg of 100 ea

O.D. × H

23 mm × 85 mm

volume

22 mL

fitting

thread for 20-400

Looking for similar products? Visit Product Comparison Guide

Application

It was used in decanting serum for residue analysis using GC/MS/MS. It was also used as reaction and development chamber in examining reaction of dioxane solution of oxalyl chloride with an electropositive metal such as Zn and Mg.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zn-Mediated Reduction of Oxalyl Chloride Forming CO and Its Application in Carbonylation Reactions.
Markovic?, Martin, et al.
Organic Letters, 17.22, 5618-5621 (2015)
Human maternal and umbilical cord blood concentrations of polybrominated diphenyl ethers.
Foster, Warren G., et al.
Chemosphere, 84.10, 1301-1309 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service