Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

14567

Sigma-Aldrich

α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol

Mr 3,000, cross-linking reagent amine reactive, carboxylic acid

Synonym(s):

O,O′-Bis[2-(succinylamino)ethyl]polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2CONH(CH2CH2O)nCH2CH2NHCOCH2CH2COOH
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

Product Name

α,ω-Bis{2-[(3-carboxy-1-oxopropyl)amino]ethyl}polyethylene glycol, Mr 3000

mol wt

Mr 3000

Quality Level

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

Ω-end

carboxylic acid

α-end

carboxylic acid

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

O(COCCOCCC(=O)O)CCOCCC(=O)O

InChI

1S/C11H20O8/c12-10(13)1-3-16-5-7-18-9-19-8-6-17-4-2-11(14)15/h1-9H2,(H,12,13)(H,14,15)

InChI key

LQRCZNATFRANRS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service