Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20557

Supelco

ORBO 1000 Precleaned Small PUF Cartridge

O.D. × L 22 mm × 7.6 cm, pkg of 3 ea

Synonym(s):

Small PUF Cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

polyurethane (foam)

Quality Level

Agency

ASTM® D4861,D4947
EPA IP-8,TO-10A

description

Small PUF
assembled

product line

ORBO

feature

precleaned

packaging

pkg of 3 ea

manufacturer/tradename

ORBO 1000

technique(s)

active air sampling: suitable

O.D. × L

22 mm × 7.6 cm

matrix

PUF polyurethane foam

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

For ORBO PUF cartridge, several US EPA and ASTM methods require a polyurethane foam (PUF) adsorbent cartridge for monitoring semivolatiles in stack, ambient, indoor, and workplace atmospheres. A low pressure drop across the cartridge facilitates high volume sampling. ORBO-1000 small PUF cartridges are useful for ASTM D4861, EPA IP-8 and EPA TO-10A (Pesticides/PCBs); ASTM D4947 (Chlordane/Heptachlor). Small PUF cartridges are useful for the 1-5 L/min sampling range.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Felipe Faix Barby et al.
Scientific reports, 9(1), 1112-1112 (2019-02-06)
Notopteridae (Teleostei, Osteoglossiformes) represents an old fish lineage with ten currently recognized species distributed in African and Southeastern Asian rivers. Their karyotype structures and diploid numbers remained conserved over long evolutionary periods, since African and Asian lineages diverged approximately 120

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service