Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

92-0201

US EPA Method TO-10/IP-8 and ASTM® D4861: GC Analysis of Pesticides on SPB®-608 after Collection/Desorption using ORBO-1000

suitable for GC, application for air monitoring

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

gas chromatography (GC): suitable

test parameters

sample/matrix: polyurethane foam (PUF) spiked with 250 ng each pesticide
adsorbent tube: ORBO-1000 (20557)
column: SPB-608, 30 m × 0.53 mm I.D., 0.5 μm film (25312)
oven: 150 °C (1 min) to 270 °C at 4 °C/min
detector: ECD
carrier gas: helium, 7 mL/min
injection: 1 μL

suitability

application for air monitoring

application(s)

air monitoring
environmental
industrial hygiene

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    trans-Nonachlor
    Pentachlorobenzene
    Captan
    Methoxychlor
    2,4-D methyl ester
    Heptachlor
    Octachloronaphthalene
    Mirex
    2,4,5-Trichlorophenol
    Ronnel, analytical standard
    γ-BHC, certified reference material, 1000 μg/mL in methanol
    α-BHC
    1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene
    4,4′-DDT
    Heptachlor epoxide, PESTANAL®, analytical standard, isomer B
    Pentachlorophenol
    Dieldrin
    Chlorothalonil
    Hexachlorobenzene
    Aldrin
    β-BHC
    See All (21)

analytical column

related product


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service