Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

20037

Supelco

ORBO 2000 Precleaned Large PUF Cartridge

O.D. × L 6 cm × 7.6 cm, pkg of 1 ea

Synonym(s):

Large PUF Cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

polyurethane (foam)

Quality Level

Agency

ASTM® D6209
EPA IP-7,TO-13A,TO-4A,TO-9A

description

Large PUF
assembled

product line

ORBO

feature

precleaned

packaging

pkg of 1 ea

manufacturer/tradename

ORBO 2000

technique(s)

active air sampling: suitable

O.D. × L

6 cm × 7.6 cm

matrix

PUF polyurethane foam

application(s)

air monitoring
environmental
industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

  • ORBO 2000 precleaned large polyurethane foam (PUF) cartridge exhibit a low-pressure drop across the cartridge that facilitates high volume sampling.
  • PUF adsorbent cartridges are useful for ASTM D6209, EPA IP-7 and EPA TO-13 (PAHs); EPA TO-4A (Pesticides/PCBs); EPA TO-9A (Dioxins) methods in monitoring semivolatiles in stack, ambient, indoor and workplace atmospheres.

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
ORBO is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service