Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z744100

CoolSafe system

to hold, 96 x 0.2mL tubes

Synonym(s):

Cooling Box, cooling block, dry block, tube cooling block

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41123300

feature

to hold 96 x 0.2mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Diversified Biotech CSAF-1000

Looking for similar products? Visit Product Comparison Guide

General description

System Includes: 1 CoolSafe Box with Lid, 1 CHAM-1000, One 1ºC and one -10ºC Cool Bricks. Versatile - Designed for 0.2ml tubes (strips or trays), 96-well and 384-well microplates, 0.5ml, 1.5ml, 5ml tubes. Specially Formulated - Cool Bricks maintain a constant temperature (1°C or –10°C) for up to 10 hours.

Application

PCR

Features and Benefits

Cool Bricks maintain a constant temperature (1°C or –10°C) for up to 10 hours.

Legal Information

CoolSafe is a trademark of Diversified Biotech, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service