Z742585
CoolCube™ Microtube and PCR Plate cooler
Synonym(s):
PCR plate cooler
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
packaging
pkg of 1
manufacturer/tradename
Benchmark Scientific R1000
technique(s)
PCR: suitable
Related Categories
General description
The CoolCube is designed to store samples safely on the
lab bench without sample degradation due to temperature
increase or fluctuation. Simply place the CoolCube in a freezer
overnight and it′s ready for use, keeping samples cold, but not frozen
maintaining temperature at approximately 0° C for up to four hours.
1.5 & 2.0ml microtubes
0.2ml PCR tubes & strips
96 well PCR plates
lab bench without sample degradation due to temperature
increase or fluctuation. Simply place the CoolCube in a freezer
overnight and it′s ready for use, keeping samples cold, but not frozen
maintaining temperature at approximately 0° C for up to four hours.
- Maintains 0°C for up to 4 hours
- Also for use as a PCR workstation
- Two sides, ideal for:
1.5 & 2.0ml microtubes
0.2ml PCR tubes & strips
96 well PCR plates
Legal Information
CoolCube is a trademark of Benchmark Scientific, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service