Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

BR781260

BRAND® PCR Mini-cooler with transparent lid

Synonym(s):

Accessories, Mini cooler, PCR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103025
NACRES:
NB.51

material

polypropylene

packaging

pack of 2 ea

manufacturer/tradename

BRAND 781260

technique(s)

PCR: suitable

Related Categories

General description

For protecting samples from warming. The mini PCR cooler keeps samples at 4 °C for approx. 3 hours. The insulating gel changes from violet to pink at 7 °C. Suitable for 0.2 mL single vessels, 8-strips, 12-strips as well as 96-well PCR plates.

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service