Key Documents
CLS432070
Corning® CoolSink®
XT96F, flat bottom plate module
Synonym(s):
Flow indicator
About This Item
UNSPSC Code:
41103025
NACRES:
NC.01
feature
flat bottom plate module
packaging
pkg of 1
manufacturer/tradename
Corning 432070
L × W × H
12.8 cm × 8.5 cm × 1.7 cm
Related Categories
General description
Corning CoolSink thermo-conductive plate and reservoir modules provide uniform temperature to all wells, regardless of position. When placed onto a temperature source such as ice, dry ice, liquid nitrogen or water baths, the CoolSink module will rapidly adapt to that temperature - from -196°C to >+100°C.
  • CoolSink modules ensure temperature uniformity when cooling, snap freezing, heating or thawing samples.
  • All CoolSink modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
  • All modules are compatible with all temperature sources.
  • For 55 mL reagent reservoirs
Legal Information
CoolSink is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)