Key Documents
Z743322
Brayer for Breathe-EASY®) sealing membrane, 4 in firm deluxe rubber roller
pack of 1 ea
About This Item
UNSPSC Code:
41122410
NACRES:
NB.35
material
rubber (soft)
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech BRAY-5000
General description
Handy 4 inch soft rubber brayer ensures a good seal between microplate and sealing films.
Legal Information
Breathe-Easy is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)