Key Documents
Z380067
Sigma-Aldrich
Brayer for Breathe-Easy® sealing membrane
4" soft rubber roller
Synonym(s):
breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture
About This Item
UNSPSC Code:
41122410
NACRES:
NB.22
material
rubber (soft)
sterility
non-sterile
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech BRAY-4000
General description
Soft rubber roller (4") assures adhesion of Breathe-Easy membrane and facilitates removal of top protective sheet.
Legal Information
Breathe-Easy is a registered trademark of Diversified Biotech
Choose from one of the most recent versions:
Certificates of Analysis (COA)