Z727059
Roll-Seal™ sealing films
AeraSeal™ film, suitable for (for cell and tissue culture)
Synonym(s):
microplate sealing film, multiwell plate sealing film, sealing film
About This Item
Recommended Products
material
Rayon film
description
adhesive sealing film on rolls for high-throughput applications on automated microplate sealers
sterility
non-sterile
feature
piercable
packaging
roll of 1
manufacturer/tradename
Excel Scientific RB-81x100
W × L
81 mm × 100 m
suitability
suitable for (for cell and tissue culture)
Related Categories
General description
- Fast, efficient, high-throughput sealing; 2.5-5 seconds/plate; no handling and backing removal from individual sheets
- Hands-free; reduces chance of mishap or contamination; more uniform sealing from plate-to-plate
- Heat-free; no high temperatures to damage sensitive samples or warp plates and cause robotic handling problems
- Especially convenient when working with foils; avoids curling that occurs with individual sheets
- Economical; lower cost per plate than individual sheet seals or heat-seal films
- Easier peel for removal compared to many heat-seal films
- High-precision dispensing with automated sealers; consistent positioning on plates
- Compatible with most current adhesive SBS plate sealers
- Static-free manufacturing and packaging facilitates alignment and adhesion
- Protective packaging prevents damage during shipping
- Plastic cores eliminate corrugate fiber dust
Roll-Seal adhesive seals are currently available in three sealing materials:
ThermalSeal RTS high-vapor-barrier, clear polyolefin with ultra-strong pressure-sensitive silicone adhesive for qPCR, sitting-drop protein crystallization, HTS, storage and general incubation applications
AlumaSeal pierceable aluminum foil with pressure-sensitive acrylic adhesive for PCR, HTS and cold storage
AeraSeal breathable rayon films for cell and tissue culture, easily pierceable for sample recovery
"ThermalSeal" and "AlumaSeal" are registered trademarks. "Roll-Seal", "ThermalSeal RTS", and "AeraSeal" are trademarks of Excel Scientific, Inc.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)