Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

A9224

AeraSeal film

sterile

Synonym(s):

breathable plate seal, permeable rayon film, piercable plate seal, tissue culture plate seal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NB.22

material

Rayon film

sterility

sterile

feature

piercable

packaging

pack of 50 ea (one tamper evident zippered bag of 50)

manufacturer/tradename

Excel Scientific BS-25

technique(s)

tissue culture: suitable

L

142.9 mm

W × L

8.26 cm × 14.29 cm

W

82.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

A 140 μm thick hydrophobic porous sealing film with medical-grade adhesive for tissue culture plates, bio-blocks, and 96-well plates where gas exchange is necessary for cell or bacterial growth. AeraSeal sealing films minimize cross-contamination, spillage and evaporation. They allow uniform air and CO2 exchange for all wells, unlike plate lids which favor exchange for wells near the plate edges. Dimensions: 82.6 by 142.9 mm for standard-size tissue culture plate
  • non-cytotoxic, highly gas permeable
  • easily pierceable with pipet tips or pipets for sample recovery
  • recommended for temperatures from -20 °C to +80 °C


Note: AeraSeal films are suitable for hypoxic culture conditions.

Application

  • AeraSeal films have been used for the covering of 96-well opaque white plates to incubate bacterial cells.
  • It has been used for the sealing of 96 deep-well plate for culturing of yeast cells.
  • It has been used for the sealing of 96-well microtitre plate to separate mites.

Features and Benefits

AeraSeal film is:
  • Sterile, non-cytotoxic and highly gas permeable for widest range of cultures
  • Easily pierceable for sample recovery with pipette tips or pipettes
  • Can be used from incubator to freezer - withstands temperatures from −20 °C to +80 °C

Legal Information

AeraSeal is a trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin Hsia et al.
PloS one, 11(2), e0149483-e0149483 (2016-02-18)
Synthetic zinc finger proteins (ZFPs) can be created to target promoter DNA sequences, repressing transcription. The binding of small RNA (sRNA) to ZFP mRNA creates an ultrasensitive response to generate higher effective Hill coefficients. Here we combined three "off the
Jose Francisco Lima-Barbero et al.
Vaccines, 7(3) (2019-09-25)
The poultry red mite (PRM), Dermanyssusgallinae, is a hematophagous ectoparasite of birds with worldwide distribution that causes economic losses in the egg-production sector of the poultry industry. Traditional control methods, mainly based on acaricides, have been only partially successful, and
Allison J Lopatkin et al.
Nature communications, 8(1), 1689-1689 (2017-11-23)
In the absence of antibiotic-mediated selection, sensitive bacteria are expected to displace their resistant counterparts if resistance genes are costly. However, many resistance genes persist for long periods in the absence of antibiotics. Horizontal gene transfer (primarily conjugation) could explain
Michael J Tessaro et al.
Applied and environmental microbiology, 78(2), 604-606 (2011-11-15)
A whole-cell biosensor for glutamine (GlnLux) was constructed by transforming an Escherichia coli glutamine (Gln) auxotroph with a constitutive lux reporter gene. Measurements of Gln in plant extracts using GlnLux correlated with quantification using high-performance liquid chromatography (Spearman's r =
Keara Lane et al.
Cell systems, 8(3), 183-196 (2019-03-25)
During an infection, immune cells must identify the specific level of threat posed by a given bacterial input in order to generate an appropriate response. Given that they use a general non-self-recognition system, known as Toll-like receptors (TLRs), to detect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service