Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z719137

Wash Bottle

capacity 500 mL (16 oz)

Synonym(s):

bottle lab, wash bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.17

material

low-density polyethylene
polypropylene cap
polypropylene screw closure
red bottle
wide-mouth bottle

Joint

Joints threaded neck (38)

feature

label Distilled water
vented
with cap

packaging

pack of 4 ea

manufacturer/tradename

Nalgene 2402-0500

cap diam.

38 mm

capacity

500 mL (16 oz)

General description

  • Nalgene® wash bottles are manufactured from non-toxic, high-quality, laboratory-grade low-density polyethylene (LDPE).
  • A wide-mouth facilitates faster, easier filling.
  • Chemically resistant bottles and tubulation are molded in one piece for uniform flow.
  • Contents can be dispensed without tipping or shaking.
  • The dispensing tip can be cut back to increase flow.
  • Includes a polypropylene (PP) leakproof screw closure for sample protection.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service