Key Documents
Z177024
LDPE wash bottles
capacity 500 mL, Wide-mouth
Synonym(s):
bottle lab, laboratory bottles, wash bottle
About This Item
UNSPSC Code:
41103209
NACRES:
NB.17
material
low-density polyethylene bottle
natural bottle
polypropylene cap
wide-mouth bottle
Joint
Joints threaded neck (53)
feature
label no label
vented
with cap
packaging
pack of 6 ea
manufacturer/tradename
Bel-Art F11620-0500
capacity
500 mL
General description
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.
Choose from one of the most recent versions:
Certificates of Analysis (COA)