Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

Z177032

LDPE wash bottles

Wide-mouth, capacity 1,000 mL

Synonym(s):

bottle lab, laboratory bottles, wash bottle

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103209
NACRES:
NB.17

material

low-density polyethylene bottle
polypropylene cap
round bottle
white bottle
wide-mouth bottle

Joint

Joints threaded neck (53)

Agency

suitable for EPA OTM-45

feature

label no label
vented
with cap

packaging

pack of 3 ea

manufacturer/tradename

Bel-Art F11620-1000

capacity

1,000 mL

application(s)

PFAS testing

Looking for similar products? Visit Product Comparison Guide

General description

Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service