Documents
Z104647
Sigma-Aldrich
LDPE wash bottles
Narrow-mouth, capacity 500 mL
Synonym(s):
bottle lab, laboratory bottles, wash bottle
About This Item
UNSPSC Code:
41103209
NACRES:
NB.17
material
low-density polyethylene bottle
narrow-mouth bottle
natural bottle
polypropylene cap
Joint
Joints threaded neck (38)
feature
label no label
vented
with cap
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F11618-0016
capacity
500 mL
General description
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.