Key Documents
Z380334
Jars Nalgene® style 2117
capacity 1000 mL
Synonym(s):
laboratory jars, nalgene jars
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
Pricing and availability is not currently available.
material
polymethylpentene bottle
polypropylene cap
polypropylene screw closure
round transparent bottle
Joint
Joints threaded neck (120)
description
Autoclavable with closure completely unscrewed
feature
with cap
packaging
pkg of 4
manufacturer/tradename
Nalgene® 2117-1000
bottle diam. × H
112 mm × 151 mm
capacity
1000 mL
General description
Transparent, with good chemical resistance.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)