Skip to Content
MilliporeSigma
All Photos(1)

Documents

B9407

Sigma-Aldrich

Nalgene® bottles, style 2104

capacity 1,000 mL

Synonym(s):

HDPE bottles, nalgene bottle, nalgene bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

polypropylene cap
polypropylene screw closure
round bottle
translucent high-density polyethylene bottle

Joint

Joints threaded neck (63)

Agency

suitable for EPA OTM-45

description

Not autoclavable

feature

with cap

packaging

pack of 6 ea

manufacturer/tradename

Nalgene® 2104-0032

bottle diam. × H

91 mm × 199 mm

capacity

1,000 mL

Looking for similar products? Visit Product Comparison Guide

General description

Boston-style (round) wide-mouth bottles

Features and Benefits

  • Nalgene® bottles are ideal for storing chemicals, materials, and specimens.
  • Laboratory-grade HDPE material for reliably low leachables and extractables.
  • Suitable for use in freezers (up to -100°C).
  • Rigid, uniform walls offer protection against splitting or punctures.
  • Bottles have excellent durability and are chemically resistant to most acids, bases, and alcohols.
  • Linerless caps ensure a leak-proof seal for sample protection.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service