Documents
Z353744
Sigma-Aldrich
Nalgene® centrifuge bottles, wide-mouth with sealing caps Style 3141
nominal capacity 500 mL
Synonym(s):
leak proof centrifuge bottle
About This Item
UNSPSC Code:
41121703
NACRES:
NB.11
material
flat bottom bottle
polypropylene copolymer (with sealing closure)
round bottle
translucent bottle
Joint
Joints threaded neck (63)
feature
RCF 13,700 × g
with cap
packaging
pack of 4 ea
manufacturer/tradename
Nalgene® 3141-0500
parameter
13700 × g max. RCF
bottle diam. × L
69.5 mm × 159.3 mm
nominal capacity
500 mL
General description
Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with these PPCO centrifuge bottles, which include a sealing closure for added leakproof assurance at maximum speeds when spinning hazardous materials. Molded of rigid and robust polypropylene copolymer, these translucent bottles are designed for pelleting applications, such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations. Autoclavable with excellent mechanical strength.
Features and Benefits
  • Sturdy polypropylene copolymer (PPCO).
  • Designed for low- to moderate-centrifugation speed, large-volume, biological and chemical samples.
  • Ideal for use in cell culture harvesting and protein biochemistry applications, including ammonium sulfate precipitations.
  • Features a leakproof sealing closure for sample protection.
  • Autoclave safe.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries