Key Documents
Z364517
Sigma-Aldrich
Nalgene® media bottles, style 2019
capacity 500 mL
Synonym(s):
bottle nalgene, nalgene containers, sterile bottles
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
material
high-density polyethylene cap
square bottle
transparent PETG
Joint
Joints threaded neck (38-430mm)
description
Not autoclavable
sterility
sterile; γ-irradiated
feature
graduations
with cap
packaging
pack of 24 ea (2 xpack of 12)
manufacturer/tradename
Nalgene® 2019-0500
capacity
500 mL
General description
  • Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
  • Designed for storing buffers, cultures, and culture media for various laboratory applications.
  • Features a leakproof closure for sample protection.
  • Heat-shrink band around closure and neck facilitate tamper-evident seal.
  • Ideal for refrigerated/frozen storage down to -40°C.
Media bottles (square), with white HDPE screw closure. Low gas permeability, non-pyrogenic. Heat-shrink band around closure and neck provides tamper-evident seal.
Features and Benefits
  • Avoids depyrogenation, expensive washing, and autoclaving steps
  • Sterile and non-pyrogenic
  • Obtained in low particulate format
  • Validated by extensive documentation
  • Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
  • Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)