Key Documents
Z333999
BRAND® pipette fillers
universal design, for pipettes up to 100 mL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
material
natural rubber
description
3 valves
feature
for pipettes up to 100 mL
manufacturer/tradename
BRAND 25315
volume
100 mL
General description
Simple pipetting aids for volumetric and graduated pipettes. Control of the functions by squeezing the appropriate valves between thumb and fore finger.
Other Notes
BR25315 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)