Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

HS20623

Safety Bulb Pipette Filler

capacity 15 mL, red

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

natural rubber
red

packaging

pack of 3 ea

manufacturer/tradename

Heathrow Scientific HS20623

L × O.D. × diam.

106 mm × 41 mm × 4.5-6.5 mm, (hole diam)

capacity

15 mL

General description

The Safety Bulb Pipette Filler, made of natural rubber, features a white ABS attachment containing a silicone conical adapter that seals around pipettes without twisting.

Features and Benefits

  • Pipette does not contact hard plastic, which reduces the risk of cracking and chipping
  • 15 mL volume, 4.2 in L x 1.6 in outer dia. x 0.2-0.3 in hole dia. (106 mm L x 41 mm OD x 4.5-6.5 mm hole dia.)
  • Elastic band for hanging on reagent bottle or wrist
  • Filler fits standard size pipettes

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service