Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P7924

Sigma-Aldrich

Pipette Pump II

green

Synonym(s):

pipette filler, thumb wheel pipette pump

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

material

plastic
silicone

packaging

pack of 1 ea

manufacturer/tradename

Bel-Art F37911-1010

volume

10 mL

color

green

Looking for similar products? Visit Product Comparison Guide

General description

For safe and accurate pipetting with one-handed operation. Rotation of the thumb wheel provides precise control of filling or dispensing. Rapid emptying is possible by pressing the plunger or the quick release bar (not available on the 0.2 mL pump). Color coded for easy identification.

Compatibility

for pipettes up to 10 mL

Legal Information

Pipette Pump is a trademark of SP Scienceware, a division of SP

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service