Skip to Content
MilliporeSigma
All Photos(1)

Documents

TRUEENZ

Sigma-Aldrich

TrueGel3D Enzymatic Cell Recovery Solution

solution, 500 μl

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

shipped in

dry ice

storage temp.

−70°C

General description

TrueGel3D enzymatic cell recovery solution is extracted from Chaetomium gracile, which cleave glycosidic bonds of dextran polymers. It is used to dissolve dextran polymer to recover chemically fixed or live cells for post-culture analyses including RT-PCR, western blots and other molecular biological studies.

Application

TrueGel3D enzymatic cell recovery solution is used to dissolve dextran-based hydrogels to recover cells for post-culture analysis

Components

TRUEENZ-500UL contains TrueGel3D enzymatic cell recovery solution (dextranase)

Caution

Avoid loss of enzymatic activity by storing at -70°C

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

A specific protocol for TrueGel3D™ hydrogel CD cell-degradable crosslinker and RGD peptide (TRUE1) kits for 3D cell culture is available

A specific protocol for TrueGel3D™ hydrogel CD cell-degradable crosslinker and RGD peptide (TRUE1) kits for 3D cell culture is available

A specific protocol for TrueGel3D™ hydrogel CD cell-degradable crosslinker and RGD peptide (TRUE1) kits for 3D cell culture is available

A specific protocol for TrueGel3D™ hydrogel CD cell-degradable crosslinker and RGD peptide (TRUE1) kits for 3D cell culture is available

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service