Skip to Content
MilliporeSigma

TRUETHIO

TrueGel3D Thioglycerol Solution

solution, 180 μl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Form:
liquid
Technique(s):
cell attachment: suitable
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

technique(s)

cell attachment: suitable

shipped in

dry ice

storage temp.

−20°C

General description

The thiol group of thioglycerol covalently binds to SH-reactive groups of polymers (Dextrans/PVA) to modulate gel strength.

Application

TrueGel3D thiglycerol is used as negative control in place of bioactive peptides to modify hydrogels.

Other Notes

Excess use of thioglycerol results in few thiol-reactive groups for crosslinking
TRUETHIO-180UL contains thioglycerol


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library