Skip to Content
MilliporeSigma

TRUECD

TrueGel3D Crosslinker

CD cell-degradable crosslinker

Synonym(s):

Crosslinker

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.52
UNSPSC Code:
12352207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

storage temp.

−20°C

General description

CD cell-degradable crosslinker is comprised of conjugated peptide along with polyethylene glycol and thiol (SH) groups. The thiol groups form stable thioether bond with thiol-reactive groups of the polymers (Mal-PVA, Mal-Dextran, SG-PVA and SG-Dextran) and results in a stable hydrogel.

The conjugated peptide has matrix metalloprotease (MMP)-cleavage sites (Pro-Leu-Gly-Leu-Trp-Ala), which allows matrix metalloproteases (MMP1, MMP3, MMP7 and MMP9) to locally degrade the polymer network. It allows cells to spread and migrate within the gel if they express the indicated MMPs.

Application

TRUECD is used to crosslink PVA and dextran polymers to generate cell-degradable hydrogel

Features and Benefits

MMP degradable

Other Notes

TRUECD contains CD cell-degradable crosslinker lyophilized and water.

Disclaimer

Avoid prolong exposure of CD cell-degradable crosslinker to air


Still not finding the right product?

Explore all of our products under TrueGel3D Crosslinker


Storage Class

10 - Combustible liquids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library