Skip to Content
MilliporeSigma

TRUE2

TrueGel3D Hydrogel Kits

FAST-DEXTRAN, allow cell recovery, PEG non cell-degradable crosslinker

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Storage temp.:
−70°C
Shipped in:
dry ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

General description

The gel is formed by stable thioether bonds between maleimide groups of dextran (FAST-DEXTRAN) and thiol groups on PEG non cell-degradable crosslinker. Maleimide-functionalized dextran is specifically used to decrease the time for gel formation. The dextran polymer does not have cell attachment sites, but it can be customized to conjugate RGD peptide using TrueGel3D RGD (TRUERGD-1UM) prior to the crosslinking step.

TrueGel3D enzymatic cell recovery solution (TRUEENZ-500UL) is used to dissolve dextran polymers for easy recovery of chemically fixed or live cells for post-culture analyses including RT-PCR, western blots and other molecular biological studies.

Application

TRUE2-1KT is used to grow cells that don′t attach to hydrogel for survival but rather use their secreted extracellular matrix. It is used to immobilize cells or specimens for imaging and for bioprinting.

Features and Benefits

  • Fast gelling (takes few seconds to minutes after addition of crosslinker)
  • Allow cell recovery and post-culture analyses
  • Flexibility in modulating gel stiffness
  • Cells do not attach to hydrogel (if not customized with TrueGel3D RGD peptide)
  • Does not allow cell migration

Other Notes

TRUE2-1KT contains lyophilized FAST-DEXTRAN (polymer) and PEG non cell-degradable crosslinker with reconstitution buffer, TrueGel3D thioglycerol, and water.

Disclaimer

Avoid prolong exposure of PEG non cell-degradable crosslinker to air.

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Global Trade Item Number

SKUGTIN
TRUE2-1KT04061835539253

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service