Skip to Content
MilliporeSigma
All Photos(2)

Documents

T7407

Sigma-Aldrich

Trigalacturonic acid

≥90% (HPLC)

Synonym(s):

α-D-GalA-(1→4)-α-D-GalA-(1→4)-α-D-GalA, α-D-GalA-(1→4)-α-D-GalA-(1→4)-D-GalA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26O19
CAS Number:
Molecular Weight:
546.39
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥90% (HPLC)

form

powder

color

white

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

OC(C=O)C(O)C(OC1OC(C(OC2OC(C(O)C(O)C2O)C(O)=O)C(O)C1O)C(O)=O)C(O)C(O)=O

InChI

1S/C18H26O19/c19-1-2(20)3(21)10(9(27)14(28)29)34-18-8(26)6(24)11(13(37-18)16(32)33)35-17-7(25)4(22)5(23)12(36-17)15(30)31/h1-13,17-18,20-27H,(H,28,29)(H,30,31)(H,32,33)

InChI key

FMNDXLWVYMQMHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon J Charnock et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(19), 12067-12072 (2002-09-11)
Enzyme-catalyzed beta-elimination of sugar uronic acids, exemplified by the degradation of plant cell wall pectins, plays an important role in a wide spectrum of biological processes ranging from the recycling of plant biomass through to pathogen virulence. The three-dimensional crystal
P Biely et al.
FEBS letters, 382(3), 249-255 (1996-03-18)
Endopolygalacturonases I and II (PGI and PGII) of Aspergillus niger and an exopolygalacturonase (ExoPG) of A. tubingensis were investigated to reveal the stereochemistry of their hydrolytic action. Reduced pentagalacturonic acid (pentaGalU-ol) and reduced trigalacturonic acid (triGalU-ol) were used as non-reducing
S Berensmeier et al.
Applied microbiology and biotechnology, 64(4), 560-567 (2003-12-16)
The pectate lyase gene pelA from alkaliphilic Bacillus licheniformis strain 14A was cloned and sequenced. The nucleotide sequence corresponded to an open reading frame of 1,026 bp that codes for a 39 amino acid signal peptide and a mature protein
Y Nakahara et al.
Carbohydrate research, 200, 363-375 (1990-04-25)
Syntheses of alpha-GalA-(1----4)-alpha-GalA-(1----4)-GalA, a proposed plant wound-hormone, and its stereoisomer beta-GalA-(1----4)-alpha-GalA-(1----4)-GalA are described. The key intermediates, tert-butyldiphenylsilyl O-(6-O-acetyl-2,3,4-tri-O-benzyl-alpha-D-galactopyranosyl)-(1----4)-O-(6 -O- acetyl-2,3-di-O-benzyl-alpha-D-galactopyranosyl)-(1----4)-6-O-acetyl-2,3 -di-O- benzyl-beta-D-galactopyranoside and tert-butyldiphenylsilyl O-(6-O-acetyl-2,3,4-tri-O-benzyl-beta-D-galactopyranosyl)-(1----4)-O-(6- O- acetyl-2,3-di-O-benzyl-alpha-D-galactopyranosyl)-(1----4)-6-O-acetyl-2,3 -di-O- benzyl-beta-D-galactopyranoside, were prepared by stereoselective alpha-glycosylation of tert-butyldiphenylsilyl 6-O-acetyl-2,3-di-O-benzyl-beta-D-galactopyranoside with O-(6-O-acetyl-2,3,4-tri-O-benzyl-alpha-D-galactopyranosyl)-(1----4)-6-O- acetyl-2,3-di-O-benzyl-D-galactopyranosyl
Judicaël Parisot et al.
Carbohydrate research, 337(16), 1427-1433 (2002-09-03)
A new exopolygalacturonate lyase (Pel) gene of the hyperthermophilic bacterium Thermotoga maritima was cloned and overexpressed in Escherichia coli cells. A 42 kDa monomeric Pel was shown to undergo N-terminal processing by cleavage at a putative site between alanine and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service