P7536
Pectin
for use with Total Dietary Fiber Control Kit, TDF-C10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
lyophilized powder
Quality Level
quality
for use with Total Dietary Fiber Control Kit, TDF-C10
technique(s)
photometry: suitable
application(s)
food and beverages
general analytical
storage temp.
2-8°C
General description
Pectin is a major component of primary cell walls of all land plants. It encompasses a range of galacturonic acid-rich polysaccharides, and finds applications in food, pharmaceuticals and other industries.
Application
Internal control for use with the Total Dietary Fiber Assay Kit.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Pectin: cell biology and prospects for functional analysis.
Plant Molecular Biology, 47(1-2), 9-27 (2001)
Chemistry and uses of pectin - A review
Critical Reviews in Food Science and Nutrition, 37(1), 47-73 (1997)
Plant biology (Stuttgart, Germany), 15(2), 249-263 (2012-12-21)
Pollen grains are surrounded by a sculpted wall, which protects male gametophytes from various environmental stresses and microbial attacks, and also facilitates pollination. Pollen wall development requires lipid and polysaccharide metabolism, and some key genes and proteins that participate in
Postepy biochemii, 59(1), 89-94 (2013-07-05)
Pectin represents a very complex, heterogeneous family of plant cell wall polysaccharides that play a significant role in plant growth, morphology, development, and plant defense and also serves as a gelling and stabilizing polymer in diverse food and specialty products
Food chemistry, 138(2-3), 2026-2035 (2013-02-16)
The formation of blue coloured ferric anthocyanin chelates and their colour stability during storage and thermal treatment were monitored in a pH range relevant to food (3.6-5.0). Liquid model systems were composed of different types of Citrus pectins, juices (J)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service