Skip to Content
MilliporeSigma

81325

Polygalacturonic acid

≥90% (enzymatic)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C6H8O6)n
CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥90% (enzymatic)

form

powder

mol wt

Mr 25000-50000(lit.)

color

white to light beige

mp

60  °C ((140 °F ))

storage temp.

room temp

SMILES string

O1[C@@H]([C@@H]([C@H]([C@H]([C@H]1C(=O)O)O)O)O)O

InChI

1S/C6H10O7/c7-1-2(8)4(5(10)11)13-6(12)3(1)9/h1-4,6-9,12H,(H,10,11)/t1-,2+,3+,4-,6-/m0/s1

InChI key

AEMOLEFTQBMNLQ-BKBMJHBISA-N

Application

Polygalacturonic acid may be used to identify, differentiate and characterize plant cell wall degrading polygalacturonase(s) from fungi.

Biochem/physiol Actions

In land plants, polygalacturonic acids are major components of cell wall polysaccharides (pectins). Bound polygalacturonase-inhibiting peptides (PGIP) protect pectin from degradation by fungal polygalacturonases.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.


Still not finding the right product?

Explore all of our products under Polygalacturonic acid


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library