Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3098

Sigma-Aldrich

IL-8 (77aa) (CXCL8) human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

CXCL8, NAP-1, monocyte-derived neutrophil chemotactic factor (MDNCF), neutrophil activating factor (NAF)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

8.9 kDa

packaging

pkg of 25 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL8(3576)

General description

Interleukin-8 (IL-8) is a proinflammatory CXC chemokine that can signal through the C-X-C motif chemokine receptors 1 and 2 (CXCR1 and 2). It is also called as C-X-C motif chemokine ligand 8 (CXCL8). IL-8 is secreted by monocytes and endothelial cells. The gene encoding IL-8 is localized on human chromosome 4q13.3. Recombinant human IL-8 (endothelial-derived) is an 8.9kDa protein containing 77 amino acid residues.

Application

IL-8 (77aa) (CXCL8) human has been used as a G protein-coupled receptor (GPCR) agonist.

Biochem/physiol Actions

Interleukin-8 (IL-8) chemoattracts and activates neutrophils. It is involved in guiding neutrophils and oligodendrocytes to sites of infection. The protein also stimulates angiogenesis and tumor growth. It is also associated with inflammatory diseases.

Sequence

AVLPRSAKEL RCQCIKTYSK PFHPKFIKEL RVIESGPHCA NTEIIVKLSD GRELCLDPKE NWVQRVVEKF LKRAENS

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

K Matsushima et al.
The Journal of experimental medicine, 167(6), 1883-1893 (1988-06-01)
The cDNA coding for human monocyte-derived neutrophil-specific chemotactic factor (MDNCF) was cloned from LPS-stimulated human monocyte mRNA. The cDNA sequence codes for a polypeptide consisting of 99 amino acids, including a putative signal sequence. Comparison of the deduced amino acid
Malefyt, R.
Cytokine Reference, 1061-1067 (2001)
Nathan D Camp et al.
Pharmacological research, 105, 13-21 (2016-01-17)
G protein-coupled receptors (GPCRs) are essential membrane proteins that facilitate cell-to-cell communication and co-ordinate physiological processes. At least 30 human GPCRs contain a Type I PSD-95/DLG/Zo-1 (PDZ) ligand in their distal C-terminal domain; this four amino acid motif of X-[S/T]-X-[φ]
The chemokine and chemokine receptor superfamilies and their molecular evolution.
Zlotnik A
Genome Biology, 7(12), 243-243 (2006)
Qi Sun et al.
International journal of clinical and experimental pathology, 8(7), 8376-8384 (2015-09-05)
Interleukin-8 (IL-8), matrix metalloproteinase-9 (MMP-9) and neovascularization have been implicated to be associated with biological processes, especially cancer progression. However, few studies have investigated the role of IL-8 in human hilar cholangiocarcinoma. In this study we detected the expression of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service