Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0657

Sigma-Aldrich

GROα human

≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

MGSA, Melanoma Growth Stimulating Activity

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

Quality Level

recombinant

expressed in E. coli

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

potency

1.0-4.0 ng/mL

mol wt

~7.9 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CXCL1(2919)

General description

GROα, also called CXCL1 (chemokine (C-X-C motif) ligand 1), is a member of the small soluble chemokine protein family. It belongs to the C-X-C subfamily, which is comprised of 17 members. It contains a conserved glutamic acid-leucine-arginine (ELR) domain. This protein is expressed on stromal cells such as, endothelial cells, fibroblasts and blood-circulating cells.

Biochem/physiol Actions

GROα, also called CXCL1 (chemokine (C-X-C motif) ligand 1), plays an essential role in neutrophil recruitment, and the initiation of angiogenesis. This gene is up-regulated in multiple cancer types such as, gastric cancer, renal cell carcinoma, prostate cancer and melanoma. The expression of this protein is also elevated in HER2 positive metastatic breast cancer. Its expression is increased in fibroblasts, which might be involved in breast tumor progression, and its stromal expression in breast cancer is linked to poor prognosis. This gene is also up-regulated in the aggressive phenotype of bladder cancer. Polymorphism rs4074 A in this gene is also associated with susceptibility to alcoholic cirrhosis.

Physical form

Lyophilized from a 0.2 μm filtered solution in 25% acetonitrile and 0.1% trifluoroacetic acid containing 50 μg bovine serum albumin per 1 μg of cytokine.

Analysis Note

The biological activity is measured by its ability to induce chemotaxis of mouse BaF/3 cells transfected with hCXCR2.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - (Acetonitrile)

Flash Point(C)

2.0 °C - (Acetonitrile)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Makito Miyake et al.
BMC cancer, 13, 322-322 (2013-07-03)
Chemokines, including chemokine (C-X-C motif) ligand 1 (CXCL1), may regulate tumor epithelial-stromal interactions that facilitate tumor growth and invasion. Studies have linked CXCL1 expression to gastric, colon and skin cancers, but limited studies to date have described CXCL1 protein expression
An Zou et al.
BMC cancer, 14, 781-781 (2014-10-26)
CXCL1 is a chemotactic cytokine shown to regulate breast cancer progression and chemo-resistance. However, the prognostic significance of CXCL1 expression in breast cancer has not been fully characterized. Fibroblasts are important cellular components of the breast tumor microenvironment, and recent
Hans Dieter Nischalke et al.
PloS one, 8(11), e80848-e80848 (2013-11-22)
CXCL1 (CXC chemokine-ligand-1) is a ligand for CXC chemokine receptor 2 expressed on hepatic stellate cells (HSC). Thus, CXCL1 might contribute to HSC activation and fibrogenesis. In the present study, we investigated the influence of the CXCL1 rs4074 polymorphism on
J M Schröder et al.
Journal of immunology (Baltimore, Md. : 1950), 139(10), 3474-3483 (1987-11-15)
A novel monocyte-derived neutrophil-activating peptide (MONAP) produced by lipopolysaccharide- and phorbol myristate acetate-stimulated human peripheral blood monocytes was purified by sequential ion exchange-high performance liquid chromatography (HPLC), size exclusion HPLC, and reversed phase HPLC. Biologic activities of the purified cytokine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service