Skip to Content
MilliporeSigma

F3506

N-Formyl-Met-Leu-Phe

≥97% (HPLC)

Synonym(s):

Chemotactic peptide, N-Formyl-L-methionyl-L-leucyl-L-phenylalanine, fMLF, fMLP (misnomer but widely used)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H31N3O5S
CAS Number:
Molecular Weight:
437.55
UNSPSC Code:
12352200
NACRES:
NA.32
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2315783
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97% (HPLC)

form

powder

mol wt

437.55

storage condition

(Keep container tightly closed in a dry and well-ventilated place)

technique(s)

activity assay: suitable

solubility

acetic acid: 20 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CSCC[C@H](NC=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](Cc1ccccc1)C(O)=O

InChI

1S/C21H31N3O5S/c1-14(2)11-17(23-19(26)16(22-13-25)9-10-30-3)20(27)24-18(21(28)29)12-15-7-5-4-6-8-15/h4-8,13-14,16-18H,9-12H2,1-3H3,(H,22,25)(H,23,26)(H,24,27)(H,28,29)/t16-,17-,18-/m0/s1

InChI key

PRQROPMIIGLWRP-BZSNNMDCSA-N

Gene Information

human ... FPR1(2357)

General description

N-Formyl-Met-Leu-Phe is a chemotactic peptide, which functions on polymorphonuclear leukocytes (PMN), neutrophils etc.

Research area: Cell signaling

Application

N-Formyl-Met-Leu-Phe has been used for:

  • creating gradients in Zigmond chamber chemotaxis assay performed on neutrophils
  • neutrophil chemotaxis assay
  • determination of the involvement of MAPK-activating protein kinase-2 (MAPKAPK-2) and/or p38, in the signaling pathway of human polymorphonuclear leukocytes (PMNs) stimulated by N-Formyl-Met-Leu-Phe.

Biochem/physiol Actions

N-Formyl-Met-Leu-Phe induced chemotaxis in phosphoinositide 3-kinase γ (PI3K)γ-/- neutrophils, and promotes adhesion, polymerization of F-actin, Fcγ receptor-mediated phagocytosis and intracellular Ca2+ release. It acts as an inflammatory agent and activates polymorphonuclear leukocytes (PMNs) without the formation of 5-hydroxyicosatetraenoic acid (5-HETE) and leukotriene B4 (LTB4). Its effects on neutrophils and the inhibition of adenylate cyclase are inhibited by pertussis toxin.
Potent chemotactic peptide. Induces a metabolic burst in macrophages accompanied by an increase in respiratory rate, secretion of lysosomal enzymes, and production of superoxide anion.
Potent inducer of leucocyte chemotaxis and macrophage activator. Receptors that bind formylpeptides are found on phagocytic neutrophils and have recently been identified on cells of the intestinal mucosa.


Still not finding the right product?

Explore all of our products under N-Formyl-Met-Leu-Phe


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library